1-(2-Ethylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139884-5G 139884-25G 139884-1G 139884-500MG
77631-268EA 1550.15 USD
77631-268 77653-514 77640-610 77648-606
1-(2-Ethylphenyl)-2-methylpropan-1-ol ≥95%
1-(2-Ethylphenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 178.271 g/mol
MW: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD19602036
CAS Number: 1341671-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR