1-(6-Bromo-2,3-difluorophenyl)-2-methylpropan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
139861-5G
139861-25G
139861-1G
139861-500MG
77631-620EA
1357
USD
77631-620
77657-424
77640-608
77648-604
1-(6-Bromo-2,3-difluorophenyl)-2-methylpropan-1-ol ≥95%
1-(6-Bromo-2,3-difluorophenyl)-2-methylpropan-1-ol
Precaución: For Research Use Only
Formula:
265.095 g/mol MW: C₁₀H₁₁BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD34834274 CAS Number: 1521479-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...