1-(4-Chloro-2-methoxyphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139832-5G 139832-1G 139832-25G 139832-500MG
77631-618EA 1381 USD
77631-618 77636-596 77653-512 77648-602
1-(4-Chloro-2-methoxyphenyl)-2-methylpropan-1-ol ≥95%
1-(4-Chloro-2-methoxyphenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 214.689 g/mol
MW: C₁₁H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD29959073
CAS Number: 1603321-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR