1-(5-Methoxy-2-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139807-5G 139807-25G 139807-1G 139807-500MG
77631-616EA 1206 USD
77631-616 77653-510 77640-606 77648-600
1-(5-Methoxy-2-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(5-Methoxy-2-methylphenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 194.27 g/mol
MW: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34834244
CAS Number: 2295083-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR