1-(4-Fluoro-2-methoxyphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139801-5G 139801-25G 139801-1G 139801-500MG
77631-614EA 1206 USD
77631-614 77653-508 77640-604 77648-598
1-(4-Fluoro-2-methoxyphenyl)-2-methylpropan-1-ol ≥95%
1-(4-Fluoro-2-methoxyphenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 198.234 g/mol
MW: C₁₁H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD34147405
CAS Number: 2840256-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR