1-(2,4-Dichlorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139783-5G 139783-25G 139783-1G 139783-500MG
77631-266EA 1484.23 USD
77631-266 77657-422 77640-602 77644-586
1-(2,4-Dichlorophenyl)-2-methylpropan-1-ol ≥95%
1-(2,4-Dichlorophenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 219.108 g/mol
MW: C₁₀H₁₂Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD14610243
CAS Number: 1291801-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR