1-(4-(Tert-butyl)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139753-5G 139753-1G 139753-25G 139753-500MG
77631-612EA 904 USD
77631-612 77636-594 77653-506 77644-584
1-(4-(Tert-butyl)phenyl)-2-methylpropan-1-ol ≥95%
1-(4-(Tert-butyl)phenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 206.324 g/mol
MW: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD11936105
CAS Number: 60561-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR