2-Methyl-1-(naphthalen-2-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139734-5G 139734-1G 139734-500MG 139734-25G
77631-610EA 904 USD
77631-610 77636-592 77652-346 77653-504
2-Methyl-1-(naphthalen-2-yl)propan-1-ol ≥95%
2-Methyl-1-(naphthalen-2-yl)propan-1-ol

Precaución: For Research Use Only

Formula: C₁₄H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD16847353
CAS Number: 76636-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR