1-(4-Chloro-2-fluoro-3-methylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

139730-5G 139730-1G 139730-500MG
77631-264EA 1484.23 USD
77631-264 77644-580 77644-582
1-(4-Chloro-2-fluoro-3-methylphenyl)cyclohexanol ≥95%
1-(4-Chloro-2-fluoro-3-methylphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 242.717 g/mol
MW: C₁₃H₁₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD34834231
CAS Number: 1880664-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR