8-(2,3-Dichlorophenyl)-1,4-dioxaspiro[4.5]Decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

139720-5G 139720-1G 139720-25G 139720-500MG
77631-608EA 1381 USD
77631-608 77636-590 77653-502 77644-578
8-(2,3-Dichlorophenyl)-1,4-dioxaspiro[4.5]Decan-8-ol ≥95%
8-(2,3-Dichlorophenyl)-1,4-dioxaspiro[4.5]Decan-8-ol

Precaución: For Research Use Only

Formula: 303.181 g/mol
MW: C₁₄H₁₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20862267
CAS Number: 1489712-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR