8-(2,5-Difluorophenyl)-1,4-dioxaspiro[4.5]Decan-8-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
139710-5G
139710-25G
139710-1G
139710-500MG
77631-606EA
1381
USD
77631-606
77653-500
77640-600
77644-576
8-(2,5-Difluorophenyl)-1,4-dioxaspiro[4.5]Decan-8-ol ≥95%
8-(2,5-Difluorophenyl)-1,4-dioxaspiro[4.5]Decan-8-ol
Precaución: For Research Use Only
Formula:
270.272 g/mol MW: C₁₄H₁₆F₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD21594349 CAS Number: 765913-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...