(5-Chloro-2-fluorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139694-5G 139694-1G 139694-25G 139694-500MG
77631-262EA 1560.45 USD
77631-262 77636-588 77653-498 77648-596
(5-Chloro-2-fluorophenyl)(phenyl)methanol ≥95%
(5-Chloro-2-fluorophenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 236.669 g/mol
MW: C₁₃H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD12786898
CAS Number: 1250044-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR