(4'-Chloro-[1,1'-biphenyl]-4-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139670-5G 139670-1G 139670-25G 139670-500MG
77631-604EA 1441 USD
77631-604 77636-586 77653-496 77644-574
(4'-Chloro-[1,1'-biphenyl]-4-yl)(phenyl)methanol ≥95%
(4'-Chloro-[1,1'-biphenyl]-4-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 294.775 g/mol
MW: C₁₉H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD34834194
CAS Number: 2642636-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR