(5-Fluoro-2-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139669-5G 139669-1G 139669-25G 139669-500MG
77631-260EA 1484.23 USD
77631-260 77636-584 77657-420 77648-594
(5-Fluoro-2-methylphenyl)(phenyl)methanol ≥95%
(5-Fluoro-2-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 216.251 g/mol
MW: C₁₄H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD16754527
CAS Number: 54611-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR