1,4-Dibromo-2-fluoro-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

138759-5G 138759-1G 138759-25G 138759-500MG
77631-258EA 1599.59 USD
77631-258 77636-580 77653-492 77644-570
1,4-Dibromo-2-fluoro-3-(trifluoromethyl)benzene ≥95%
1,4-Dibromo-2-fluoro-3-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 321.892 g/mol
MW: C₇H₂Br₂F₄
Storage Temperature: Ambient
MDL Number: MFCD28784693
CAS Number: 1806306-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR