2-Chloro-2,2-difluoro-1-(2-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138406-5G 138406-1G 138406-500MG
77657-418EA 2001 USD
77657-418 77640-596 77644-566
2-Chloro-2,2-difluoro-1-(2-methoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-methoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 220.6 g/mol
MW: C₉H₇ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20921137
CAS Number: 1352206-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR