4-Bromo-1-(4-fluorobenzyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

13800-5G 13800-25G 13800-1G 13800-500MG
77631-596EA 638 USD
77631-596 77653-488 77640-592 77644-562
4-Bromo-1-(4-fluorobenzyl)-1H-pyrazole ≥97%
4-Bromo-1-(4-fluorobenzyl)-1H-pyrazole

Precaución: For Research Use Only

Formula: C₁₀H₈BrFN₂
MW: 255.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06167002
CAS Number: 251925-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR