2-Cyclopropyl-5,5-dimethyl-1,3,2-dioxaborinane ≥97%

Supplier: AOBChem USA

Warning

137929-5G 137929-1G 137929-25G 137929-500MG
77631-594EA 1411 USD
77631-594 77636-574 77657-412 77648-588
2-Cyclopropyl-5,5-dimethyl-1,3,2-dioxaborinane ≥97%
2-Cyclopropyl-5,5-dimethyl-1,3,2-dioxaborinane

Precaución: For Research Use Only

Formula: 154.015 g/mol
MW: C₈H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD06801703
CAS Number: 941320-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR