1-Bromo-4-(3,3,3-trifluoroprop-1-en-2-yl)benzene ≥95%

Supplier: AOBChem USA

Warning

137645-1G 137645-5G 137645-500MG
77636-572EA 518 USD
77636-572 77657-410 77648-584
1-Bromo-4-(3,3,3-trifluoroprop-1-en-2-yl)benzene ≥95%
1-Bromo-4-(3,3,3-trifluoroprop-1-en-2-yl)benzene

Precaución: For Research Use Only

Formula: 251.0431 g/mol
MW: C₉H₆BrF₃
Storage Temperature: Ambient
MDL Number: MFCD27944312
CAS Number: 136476-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR