5-Methyl-2-furonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Methylfuran-2-carbonitrile

137584-5G 137584-25G
77634-014EA 66 USD
77634-014 77653-486
5-Methyl-2-furonitrile ≥97%
5-Methyl-2-furonitrile

Precaución: For Research Use Only

Formula: C₆H₅NO
MW: 107.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093782
CAS Number: 13714-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR