1-Bromo-2,3,5-trifluoro-4-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

13736-5G 13736-500MG 13736-25G 13736-1G
77631-248EA 1703.62 USD
77631-248 77652-342 77653-484 77640-586
1-Bromo-2,3,5-trifluoro-4-(methoxymethoxy)benzene ≥95%
1-Bromo-2,3,5-trifluoro-4-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: 271.031 g/mol
MW: C₈H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34755654
CAS Number: 851341-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR