Tert-butyl 4-chloro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

137355-5G 137355-1G 137355-25G 137355-500MG
77631-592EA 693 USD
77631-592 77636-570 77653-482 77644-556
Tert-butyl 4-chloro-2-methylbenzoate ≥97%
Tert-butyl 4-chloro-2-methylbenzoate

Precaución: For Research Use Only

Formula: 226.699 g/mol
MW: C₁₂H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30180170
CAS Number: 1872907-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR