Tert-butyl 5-fluoro-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

137348-5G 137348-25G 137348-1G 137348-500MG
77631-590EA 1034 USD
77631-590 77653-480 77640-584 77644-554
Tert-butyl 5-fluoro-2-methylbenzoate ≥95%
Tert-butyl 5-fluoro-2-methylbenzoate

Precaución: For Research Use Only

Formula: 210.245 g/mol
MW: C₁₂H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD19388679
CAS Number: 1410568-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR