4-(Tert-butyl)-3-chlorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

135798-1G 135798-5G 135798-500MG
77634-010EA 783 USD
77634-010 77653-470 77648-578
4-(Tert-butyl)-3-chlorobenzaldehyde ≥95%
4-(Tert-butyl)-3-chlorobenzaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD20483890
CAS Number: 40891-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR