5-Methyl-6-(4-morpholinyl)pyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

13481-5G 13481-1G 13481-25G 13481-500MG
77631-580EA 702 USD
77631-580 77636-562 77657-406 77644-548
5-Methyl-6-(4-morpholinyl)pyridine-3-boronic acid pinacol ester ≥97%
5-Methyl-6-(4-morpholinyl)pyridine-3-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₆H₂₅BN₂O₃
MDL Number: MFCD09037483
CAS Number: 1073372-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR