2,4-Difluoro-1-((2-methylbenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

134737-5G 134737-25G 134737-1G 134737-500MG
77631-244EA 1663.45 USD
77631-244 77657-404 77640-578 77644-546
2,4-Difluoro-1-((2-methylbenzyl)oxy)benzene ≥97%
2,4-Difluoro-1-((2-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 234.241 g/mol
MW: C₁₄H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34831153
CAS Number: 2921892-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR