1-(4-(Dimethylamino)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

134613-5G 134613-1G 134613-25G 134613-500MG
77631-242EA 1515.13 USD
77631-242 77636-560 77657-402 77644-544
1-(4-(Dimethylamino)phenyl)-2-methylpropan-1-ol ≥95%
1-(4-(Dimethylamino)phenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 193.2853 g/mol
MW: C₁₂H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD00207902
CAS Number: 4278-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR