1-Chloro-3-fluoro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

133826-5G 133826-25G 133826-1G 133826-500MG
77634-008EA 372 USD
77634-008 77657-400 77640-576 77648-574
1-Chloro-3-fluoro-2-isopropoxybenzene ≥97%
1-Chloro-3-fluoro-2-isopropoxybenzene

Precaución: For Research Use Only

Formula: 188.627 g/mol
MW: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD23114123
CAS Number: 1369889-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR