3,4-Dihydro-2H-benzo[b][1,4]dioxepine-6-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

133664-5G 133664-1G 133664-25G 133664-500MG
77631-578EA 810 USD
77631-578 77636-558 77657-398 77648-572
3,4-Dihydro-2H-benzo[b][1,4]dioxepine-6-carbaldehyde ≥95%
3,4-Dihydro-2H-benzo[b][1,4]dioxepine-6-carbaldehyde

Precaución: For Research Use Only

Formula: 178.185 g/mol
MW: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD04972613
CAS Number: 209256-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR