6-Bromo-3,4-dihydro-2H-benzo[b][1,4]dioxepine ≥95%

Supplier: AOBChem USA

Warning

133658-5G 133658-1G 133658-25G 133658-500MG
77634-006EA 342 USD
77634-006 77636-556 77657-396 77644-542
6-Bromo-3,4-dihydro-2H-benzo[b][1,4]dioxepine ≥95%
6-Bromo-3,4-dihydro-2H-benzo[b][1,4]dioxepine

Precaución: For Research Use Only

Formula: 229.071 g/mol
MW: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD31653797
CAS Number: 1279822-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR