(5-Chlorobenzo[d][1,3]dioxol-4-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-Chlorobenzo[d][1,3]dioxol-4-yl)boronicacid

133585-5G 133585-25G 133585-1G 133585-500MG
77634-002EA 377 USD
77634-002 77653-462 77640-574 77648-570
(5-Chlorobenzo[d][1,3]dioxol-4-yl)boronic acid ≥95%
(5-Chlorobenzo[d][1,3]dioxol-4-yl)boronic acid

Precaución: For Research Use Only

Formula: C₇H₆BClO₄
MW: 200.384 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382028
CAS Number: 2087452-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR