(5-(Tert-butyl)-2-methylphenyl)(phenyl)methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
133561-5G
133561-1G
133561-25G
133561-500MG
77631-238EA
1831.34
USD
77631-238
77636-552
77653-458
77648-566
(5-(Tert-butyl)-2-methylphenyl)(phenyl)methanone ≥95%
(5-(Tert-butyl)-2-methylphenyl)(phenyl)methanone
Precaución: For Research Use Only
Formula:
252.351 g/mol MW: C₁₈H₂₀O Storage Temperature: Ambient |
MDL Number:
MFCD34830469 CAS Number: 42549-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...