(5-(Tert-butyl)-2-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

133559-5G 133559-25G 133559-1G 133559-500MG
77631-236EA 1583.11 USD
77631-236 77657-390 77640-570 77648-564
(5-(Tert-butyl)-2-methylphenyl)(phenyl)methanol ≥95%
(5-(Tert-butyl)-2-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 254.367 g/mol
MW: C₁₈H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD34830468
CAS Number: 2921711-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR