1-(5-(Tert-butyl)-2-methylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

133557-5G 133557-25G 133557-1G 133557-500MG
77631-234EA 1484.23 USD
77631-234 77657-388 77640-568 77648-562
1-(5-(Tert-butyl)-2-methylphenyl)cyclohexanol ≥95%
1-(5-(Tert-butyl)-2-methylphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 246.388 g/mol
MW: C₁₇H₂₆O
Storage Temperature: Ambient
MDL Number: MFCD34830467
CAS Number: 2921842-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR