1-(5-(Tert-butyl)-2-methylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

133551-5G 133551-1G 133551-500MG
77653-452EA 2053 USD
77653-452 77644-538 77648-556
1-(5-(Tert-butyl)-2-methylphenyl)propan-1-ol ≥95%
1-(5-(Tert-butyl)-2-methylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD14529994
CAS Number: 1216085-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR