2-(5-Bromo-2-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

133316-5G 133316-25G 133316-1G 133316-500MG
77631-574EA 1171 USD
77631-574 77653-450 77640-562 77648-554
2-(5-Bromo-2-methylphenyl)propan-2-ol ≥97%
2-(5-Bromo-2-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 229.1136 g/mol
MW: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD28357466
CAS Number: 1262035-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR