4-Bromo-2-methyl-1-((3-methylbenzyl)oxy)benzene ≥95%

Supplier: AOBChem USA

Warning

132860-5G 132860-25G 132860-1G 132860-500MG
77631-570EA 1034 USD
77631-570 77653-446 77640-560 77644-534
4-Bromo-2-methyl-1-((3-methylbenzyl)oxy)benzene ≥95%
4-Bromo-2-methyl-1-((3-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 291.183 g/mol
MW: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD19156069
CAS Number: 309947-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR