2,3,6-Trichloroanisole ≥95%

Supplier: AOBChem USA

Warning

132813-1G 132813-5G 132813-500MG
77634-000EA 1042 USD
77634-000 77657-384 77648-550
2,3,6-Trichloroanisole ≥95%
2,3,6-Trichloroanisole

Precaución: For Research Use Only

Formula: C₇H₅Cl₃O
MW: 211.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000548
CAS Number: 50375-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR