N-Methoxy-N-methyl-2-furancarboxamide ≥95%

Supplier: AOBChem USA

Warning

132714-5G 132714-25G
77633-998EA 117 USD
77633-998 77657-382
N-Methoxy-N-methyl-2-furancarboxamide ≥95%
N-Methoxy-N-methyl-2-furancarboxamide

Precaución: For Research Use Only

Formula: C₇H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD06804576
CAS Number: 95091-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR