4-Bromo-6-fluoro-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

132540-5G 132540-1G 132540-25G 132540-500MG
77631-568EA 1342 USD
77631-568 77636-542 77657-378 77644-530
4-Bromo-6-fluoro-1-methyl-1H-indazole ≥97%
4-Bromo-6-fluoro-1-methyl-1H-indazole

Precaución: For Research Use Only

Formula: 229.049 g/mol
MW: C₈H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD28660090
CAS Number: 1369876-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR