3-Bromo-7-chloro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

132533-5G 132533-1G 132533-25G 132533-500MG
77631-566EA 767 USD
77631-566 77636-540 77653-442 77648-544
3-Bromo-7-chloro-1H-indazole ≥95%
3-Bromo-7-chloro-1H-indazole

Precaución: For Research Use Only

Formula: C₇H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD07781601
CAS Number: 885521-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR