Tert-butyl 4-(2-ethoxyphenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

130364-5G 130364-25G 130364-1G 130364-500MG
77631-220EA 1484.23 USD
77631-220 77635-434 77640-550 77644-522
Tert-butyl 4-(2-ethoxyphenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
Tert-butyl 4-(2-ethoxyphenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For Research Use Only

Formula: 321.4112 g/mol
MW: C₁₈H₂₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD34828494
CAS Number: 2969340-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR