1-(3-Chloro-2-fluoro-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

130335-5G 130335-25G 130335-1G 130335-500MG
77631-218EA 1484.23 USD
77631-218 77657-370 77640-548 77648-540
1-(3-Chloro-2-fluoro-4-methylphenyl)ethanone ≥95%
1-(3-Chloro-2-fluoro-4-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 186.6106 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751712
CAS Number: 1804881-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR