8-(3-(Trifluoromethoxy)phenyl)-1,4-dioxaspiro[4.5]Decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129992-5G 129992-25G 129992-1G 129992-500MG
77631-216EA 1484.23 USD
77631-216 77635-432 77636-532 77648-536
8-(3-(Trifluoromethoxy)phenyl)-1,4-dioxaspiro[4.5]Decan-8-ol ≥95%
8-(3-(Trifluoromethoxy)phenyl)-1,4-dioxaspiro[4.5]Decan-8-ol

Precaución: For Research Use Only

Formula: 318.288 g/mol
MW: C₁₅H₁₇F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD34828281
CAS Number: 2921866-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR