1-(2-Fluoro-5-methoxyphenyl)cyclooctanol ≥97%

Supplier: AOBChem USA

Warning

129829-5G 129829-25G 129829-1G 129829-500MG
77631-214EA 1484.23 USD
77631-214 77653-432 77640-544 77648-534
1-(2-Fluoro-5-methoxyphenyl)cyclooctanol ≥97%
1-(2-Fluoro-5-methoxyphenyl)cyclooctanol

Precaución: For Research Use Only

Formula: C₁₅H₂₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34828247
CAS Number: 2921859-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR