1-(2-Fluoro-5-methoxyphenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

129667-5G 129667-1G 129667-25G 129667-500MG
77631-212EA 1484.23 USD
77631-212 77636-530 77657-368 77644-520
1-(2-Fluoro-5-methoxyphenyl)cycloheptanol ≥95%
1-(2-Fluoro-5-methoxyphenyl)cycloheptanol

Precaución: For Research Use Only

Formula: 238.298 g/mol
MW: C₁₄H₁₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD34828178
CAS Number: 2921849-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR