1-(2-Fluoro-3-(trifluoromethyl)phenyl)cyclooctanol ≥97%

Supplier: AOBChem USA

Warning

129585-5G 129585-25G 129585-1G 129585-500MG
77631-210EA 1484.23 USD
77631-210 77657-366 77640-542 77648-532
1-(2-Fluoro-3-(trifluoromethyl)phenyl)cyclooctanol ≥97%
1-(2-Fluoro-3-(trifluoromethyl)phenyl)cyclooctanol

Precaución: For Research Use Only

Formula: 290.296 g/mol
MW: C₁₅H₁₈F₄O
Storage Temperature: Ambient
MDL Number: MFCD34828163
CAS Number: 2918964-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR