1-(5-Chloro-2-fluorophenyl)cyclooctanol ≥95%

Supplier: AOBChem USA

Warning

129582-5G 129582-1G 129582-500MG 129582-25G
77631-208EA 1484.23 USD
77631-208 77636-528 77652-324 77657-364
1-(5-Chloro-2-fluorophenyl)cyclooctanol ≥95%
1-(5-Chloro-2-fluorophenyl)cyclooctanol

Precaución: For Research Use Only

Formula: 256.744 g/mol
MW: C₁₄H₁₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD34828162
CAS Number: 2918958-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR