1-(3-(Trifluoromethyl)phenyl)cyclooctanol ≥97%

Supplier: AOBChem USA

Warning

129549-5G 129549-1G 129549-25G 129549-500MG
77631-204EA 1777.78 USD
77631-204 77636-524 77653-428 77648-528
1-(3-(Trifluoromethyl)phenyl)cyclooctanol ≥97%
1-(3-(Trifluoromethyl)phenyl)cyclooctanol

Precaución: For Research Use Only

Formula: 272.306 g/mol
MW: C₁₅H₁₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD34828151
CAS Number: 2921707-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR