3-Bromo-4-chloro-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

129519-5G 129519-1G 129519-25G 129519-500MG
77631-560EA 1264 USD
77631-560 77636-520 77657-360 77644-516
3-Bromo-4-chloro-2-methylaniline ≥97%
3-Bromo-4-chloro-2-methylaniline

Precaución: For Research Use Only

Formula: C₇H₇BrClN
MW: 220.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34165795
CAS Number: 1936457-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR